4-(4-Methylpiperazin-1-yl)benzoyl chloride
Lieferant: Apollo Scientific
OR46673-1G
OR46673-100MG
OR46673-250MG
OR46673-500MG
OR46673-5G
APOSOR46673-1GEA
0
EUR
APOSOR46673-1G
APOSOR46673-100MG
APOSOR46673-250MG
APOSOR46673-500MG
APOSOR46673-5G
4-(4-Methylpiperazin-1-yl)benzoyl chloride
4-(4-Methylpiperazin-1-yl)benzoyl chloride
Formel:
C₁₂H₁₅ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD20483877 CAS-Nummer: 401495-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...