2-Chloro-4-hydroxybenzoyl chloride

Lieferant: Apollo Scientific

OR46675-1G
APOSOR46675-1GEA 0 EUR
APOSOR46675-1G
2-Chloro-4-hydroxybenzoyl chloride
2-Chloro-4-hydroxybenzoyl chloride
Formel: C₇H₄Cl₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13174148
CAS-Nummer: 535962-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR