2-Mercaptobenzoyl chloride

Lieferant: Apollo Scientific

OR46677-1G OR46677-250MG OR46677-500MG
APOSOR46677-1GEA 0 EUR
APOSOR46677-1G APOSOR46677-250MG APOSOR46677-500MG
2-Mercaptobenzoyl chloride
2-Mercaptobenzoyl chloride
Formel: C₇H₅ClOS
Lagertemperatur: Raumtemperatur
MDL: MFCD22054937
CAS-Nummer: 1677-25-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR