3-Acetylbenzoyl chloride
Lieferant: Apollo Scientific
OR46681-1G
OR46681-250MG
OR46681-500MG
APOSOR46681-1GEA
510
EUR
APOSOR46681-1G
APOSOR46681-250MG
APOSOR46681-500MG
3-Acetylbenzoyl chloride
3-Acetylbenzoyl chloride
Formel:
C₉H₇ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13174028 CAS-Nummer: 31076-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...