3-Acetylbenzoyl chloride

Lieferant: Apollo Scientific

OR46681-1G OR46681-250MG OR46681-500MG
APOSOR46681-1GEA 510 EUR
APOSOR46681-1G APOSOR46681-250MG APOSOR46681-500MG
3-Acetylbenzoyl chloride
3-Acetylbenzoyl chloride
Formel: C₉H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13174028
CAS-Nummer: 31076-85-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR