2-(Methylsulphonyl)benzoyl chloride

Lieferant: Apollo Scientific

OR46682-1G OR46682-250MG OR46682-500MG OR46682-2.5G
APOSOR46682-1GEA 0 EUR
APOSOR46682-1G APOSOR46682-250MG APOSOR46682-500MG APOSOR46682-2.5G
2-(Methylsulphonyl)benzoyl chloride
2-(Methylsulphonyl)benzoyl chloride
Formel: C₈H₇ClO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD12025162
CAS-Nummer: 49639-13-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR