2-(Methylthio)benzoyl chloride

Lieferant: Apollo Scientific

OR46685-1G OR46685-250MG OR46685-500MG
APOSOR46685-1GEA 418 EUR
APOSOR46685-1G APOSOR46685-250MG APOSOR46685-500MG
2-(Methylthio)benzoyl chloride
2-(Methylthio)benzoyl chloride
Formel: C₈H₇ClOS
Molecular Weight: 186.66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652997
CAS-Nummer: 1442-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR