2-(1H-Imidazol-1-yl)acetyl chloride

Lieferant: Apollo Scientific

OR46687-1G
APOSOR46687-1GEA 510 EUR
APOSOR46687-1G
2-(1H-Imidazol-1-yl)acetyl chloride
2-(1H-Imidazol-1-yl)acetyl chloride
Formel: C₅H₅ClN₂O
Molecular Weight: 144,559 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28664001
CAS-Nummer: 160975-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR