1-Pyridin-4-yl-1,2,3,4-tetrahydroisoquinoline
Lieferant: Apollo Scientific
OR4669-1G
APOSOR4669-1GEA
372
EUR
APOSOR4669-1G
1-Pyridin-4-yl-1,2,3,4-tetrahydroisoquinoline
1-Pyridin-4-yl-1,2,3,4-tetrahydroisoquinoline
Formel:
C₁₄H₁₄N₂ Molecular Weight: 210,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02323229 CAS-Nummer: 180272-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...