4-Methoxycyclohexanecarbonyl chloride

Lieferant: Apollo Scientific

OR46690-1G OR46690-250MG OR46690-500MG
APOSOR46690-1GEA 0 EUR
APOSOR46690-1G APOSOR46690-250MG APOSOR46690-500MG
4-Methoxycyclohexanecarbonyl chloride
4-Methoxycyclohexanecarbonyl chloride
Formel: C₈H₁₃ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03424700
CAS-Nummer: 123790-13-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR