Methyl (4-oxocyclohexyl)acetate

Lieferant: Apollo Scientific

OR46693-1G
APOSOR46693-1GEA 201 EUR
APOSOR46693-1G
Methyl (4-oxocyclohexyl)acetate
Methyl (4-oxocyclohexyl)acetate
Formel: C₉H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09031995
CAS-Nummer: 66405-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR