3-Chloro[1,2,4]triazolo[4,3-a]pyridine

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-[1,2,4]triazolo[4,3-a]pyridine

OR46696-1G
APOSOR46696-1GEA 102 EUR
APOSOR46696-1G
3-Chloro[1,2,4]triazolo[4,3-a]pyridine
3-Chloro[1,2,4]triazolo[4,3-a]pyridine
Formel: C₆H₄ClN₃
Molecular Weight: 153,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09901818
CAS-Nummer: 4922-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR