4-(Chloromethyl)-2-phenyl-1,3-oxazole

Lieferant: Apollo Scientific

OR46698-1G
APOSOR46698-1GEA 435 EUR
APOSOR46698-1G
4-(Chloromethyl)-2-phenyl-1,3-oxazole
4-(Chloromethyl)-2-phenyl-1,3-oxazole
Formel: C₁₀H₈ClNO
Molecular Weight: 193,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738535
CAS-Nummer: 99073-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR