Amino(4-methoxyphenyl)acetonitrile hydrochloride

Lieferant: Apollo Scientific

OR46701-1G
APOSOR46701-1GEA 451 EUR
APOSOR46701-1G
Amino(4-methoxyphenyl)acetonitrile hydrochloride
Amino(4-methoxyphenyl)acetonitrile hydrochloride
Formel: C₉H₁₀N₂O·ClH
Molecular Weight: 198,649 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27959559
CAS-Nummer: 5537-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR