2-Chloro-3-iodophenol

Lieferant: Apollo Scientific

OR46706-1G
APOSOR46706-1GEA 745 EUR
APOSOR46706-1G
2-Chloro-3-iodophenol
2-Chloro-3-iodophenol
Formel: C₆H₄ClIO
Lagertemperatur: Raumtemperatur
MDL: MFCD08166415
CAS-Nummer: 666727-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR