2-Chloro-5-phenylnicotinonitrile

Lieferant: Apollo Scientific

OR46707-1G
APOSOR46707-1GEA 261 EUR
APOSOR46707-1G
2-Chloro-5-phenylnicotinonitrile
2-Chloro-5-phenylnicotinonitrile
Formel: C₁₂H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00231610
CAS-Nummer: 10177-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR