1-Methylcyclopropanecarboxamide

Lieferant: Apollo Scientific

OR46718-1G OR46718-50MG OR46718-250MG OR46718-500MG
APOSOR46718-1GEA 589 EUR
APOSOR46718-1G APOSOR46718-50MG APOSOR46718-250MG APOSOR46718-500MG
1-Methylcyclopropanecarboxamide
1-Methylcyclopropanecarboxamide
Formel: C₅H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11845570
CAS-Nummer: 15910-91-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR