Cyclopentanecarboximidamide hydrochloride

Lieferant: Apollo Scientific

OR46719-1G OR46719-250MG OR46719-500MG OR46719-2.5G
APOSOR46719-1GEA 0 EUR
APOSOR46719-1G APOSOR46719-250MG APOSOR46719-500MG APOSOR46719-2.5G
Cyclopentanecarboximidamide hydrochloride
Cyclopentanecarboximidamide hydrochloride
Formel: C₆H₁₂N₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08752286
CAS-Nummer: 68284-02-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR