2-Aminopropanenitrile

Lieferant: Apollo Scientific

OR46724-250MG
APOSOR46724-250MGEA 304 EUR
APOSOR46724-250MG
2-Aminopropanenitrile
2-Aminopropanenitrile
Formel: C₃H₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00798450
CAS-Nummer: 2134-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR