4-(1,3-Benzothiazol-2-yl)isoxazol-5-amine

Lieferant: Apollo Scientific

Synonyme: 5-Isoxazolamine, 4-(2-benzothiazolyl)

OR46726-1G
APOSOR46726-1GEA 386 EUR
APOSOR46726-1G
4-(1,3-Benzothiazol-2-yl)isoxazol-5-amine
4-(1,3-Benzothiazol-2-yl)isoxazol-5-amine
Formel: C₁₀H₇N₃OS
Molecular Weight: 217,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656614
CAS-Nummer: 208938-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR