7-Chlor-2-mercaptobenzothiazol

Lieferant: Apollo Scientific

Synonyme: 7-chloro-3H-benzothiazole-2-thione

OR46732-1G
APOSOR46732-1GEA 72.5 EUR
APOSOR46732-1G
7-Chlor-2-mercaptobenzothiazol
7-Chloro-2-mercaptobenzothiazole
Formel: C₇H₄ClNS₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06408879
CAS-Nummer: 1849-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR