N-[4-(1,3-Benzothiazol-2-yl)phenyl]-2-(1H-1,2,3-benzotriazol 95%

Lieferant: Apollo Scientific
Achtung

OR46737-1G
APOSOR46737-1GEA 944 EUR
APOSOR46737-1G
N-[4-(1,3-Benzothiazol-2-yl)phenyl]-2-(1H-1,2,3-benzotriazol 95%
N-[4-(1,3-Benzothiazol-2-yl)phenyl]-2-(1H-1,2,3-benzotriazol
Formel: C₂₁H₁₅N₅OS
Molecular Weight: 385,442 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10209041
CAS-Nummer: 1014580-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR