4-Chloropyrrolo[1,2-b]pyridazine-3-carbonitrile

Lieferant: Apollo Scientific

OR46747-250MG
APOSOR46747-250MGEA 607 EUR
APOSOR46747-250MG
4-Chloropyrrolo[1,2-b]pyridazine-3-carbonitrile
4-Chloropyrrolo[1,2-b]pyridazine-3-carbonitrile
Formel: C₈H₄ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28053422
CAS-Nummer: 1263420-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR