4-(Dimethylamino)pyridine-2-carboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR46749-250MG
APOSOR46749-250MGEA 148 EUR
APOSOR46749-250MG
4-(Dimethylamino)pyridine-2-carboxylic acid hydrochloride
4-(Dimethylamino)pyridine-2-carboxylic acid hydrochloride
Formel: C₈H₁₀N₂O₂·ClH
Molecular Weight: 202,638 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878501
CAS-Nummer: 1176419-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR