[4-(1H-Imidazol-2-yl)phenyl]boronic acid, pinacol ester
Lieferant: Apollo Scientific
Synonyme:
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-imidazole
OR46750-250MG
APOSOR46750-250MGEA
360
EUR
APOSOR46750-250MG
[4-(1H-Imidazol-2-yl)phenyl]boronic acid, pinacol ester
[4-(1H-Imidazol-2-yl)phenyl]boronic acid, pinacol ester
Formel:
C₁₅H₁₉BN₂O₂ Molecular Weight: 270,135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18383629 CAS-Nummer: 1229584-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...