4-Aminomethyl-7-azaindole
Lieferant: Apollo Scientific
Synonyme:
(1H-Pyrrolo[2,3-b]pyridin-4-yl)methanamine
OR46751-250MG
APOSOR46751-250MGEA
79.9
EUR
APOSOR46751-250MG
4-Aminomethyl-7-azaindole
4-Aminomethyl-7-azaindole
Formel:
C₈H₉N₃ Molecular Weight: 147,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458223 CAS-Nummer: 888498-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...