N,N-Dimethyl-1-(prop-2-yn-1-yl)pyrrolidin-3-amine

Lieferant: Apollo Scientific
Gefahr

OR46767-1G
APOSOR46767-1GEA 1220 EUR
APOSOR46767-1G
N,N-Dimethyl-1-(prop-2-yn-1-yl)pyrrolidin-3-amine
N,N-Dimethyl-1-(prop-2-yn-1-yl)pyrrolidin-3-amine
Formel: C₉H₁₆N₂
Molecular Weight: 152,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18278587
CAS-Nummer: 1339671-68-9
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR