(6S)-2-Nitro-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazin-6-ol 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Nitro-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazin-6-ol

OR46771-250MG
APOSOR46771-250MGEA 137 EUR
APOSOR46771-250MG
(6S)-2-Nitro-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazin-6-ol 97%
(6S)-2-Nitro-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazin-6-ol
Formel: C₆H₇N₃O₄
Molecular Weight: 185,137 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436173
CAS-Nummer: 187235-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR