Methyl-2,5-dibrombenzoat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2,5-dibromobenzoate
OR4690-25G
APOSOR4690-25GEA
69.6
EUR
APOSOR4690-25G
Methyl-2,5-dibrombenzoat 98%
Methyl-2,5-dibrombenzoat
Formel:
C₈H₆Br₂O₂ Molecular Weight: 293,94 g/mol Schmelzpunkt: 48…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144757 CAS-Nummer: 57381-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...