2-Methyl-5-phenyl-3-furancarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-5-phenylfuran-3-carboxylic acid
OR4698-1G
APOSOR4698-1GEA
72.5
EUR
APOSOR4698-1G
2-Methyl-5-phenyl-3-furancarbonsäure
2-Methyl-5-phenyl-3-furoic acid
Formel:
C₁₂H₁₀O₃ Molecular Weight: 202,21 g/mol Schmelzpunkt: 176…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221070 CAS-Nummer: 108124-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...