endo-N,9-Dimethyl-9-azabicyclo[3.3.1]nonan-3-amine

Lieferant: Apollo Scientific

OR470007-5G
APOSOR470007-5GEA 1690 EUR
APOSOR470007-5G
endo-N,9-Dimethyl-9-azabicyclo[3.3.1]nonan-3-amine
endo-N,9-Dimethyl-9-azabicyclo[3.3.1]nonan-3-amine
Formel: C₁₀H₂₀N₂
Molecular Weight: 168,279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198905
CAS-Nummer: 123259-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR