cis-5-Ethoxycarbonyl-1H-hexahydropyrrolo[3,4-b]pyrrole
Lieferant: Apollo Scientific
OR470008-1G
APOSOR470008-1GEA
601
EUR
APOSOR470008-1G
cis-5-Ethoxycarbonyl-1H-hexahydropyrrolo[3,4-b]pyrrole
cis-5-Ethoxycarbonyl-1H-hexahydropyrrolo[3,4-b]pyrrole
Formel:
C₉H₁₆N₂O₂ Molecular Weight: 184,236 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21364417 CAS-Nummer: 894854-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...