1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine

Lieferant: Apollo Scientific

OR470011-5G
APOSOR470011-5GEA 245 EUR
APOSOR470011-5G
1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine
1-Boc-4-[3-(ethoxycarbonyl)phenyl]piperazine
Formel: C₁₈H₂₆N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD07369778
CAS-Nummer: 261925-94-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR