3,5-Dimethyl-1,2-benzenediamine

Lieferant: Apollo Scientific

OR470020-5G
APOSOR470020-5GEA 136 EUR
APOSOR470020-5G
3,5-Dimethyl-1,2-benzenediamine
3,5-Dimethyl-1,2-benzenediamine
Formel: C₈H₁₂N₂
Molecular Weight: 136.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00972722
CAS-Nummer: 3171-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR