(S)-4-Benzyl-3-morpholinecarboxylic acid ≥98%

Lieferant: Apollo Scientific

OR470023-100MG
APOSOR470023-100MGEA 0 EUR
APOSOR470023-100MG
(S)-4-Benzyl-3-morpholinecarboxylic acid ≥98%
(S)-4-Benzyl-3-morpholinecarboxylic acid
Formel: C₁₂H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06799463
CAS-Nummer: 1235011-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR