N-Isopropyl-4-nitrobenzylamine

Lieferant: Apollo Scientific

OR470027-5G
APOSOR470027-5GEA 397 EUR
APOSOR470027-5G
N-Isopropyl-4-nitrobenzylamine
N-Isopropyl-4-nitrobenzylamine
Formel: C₁₀H₁₄N₂O₂
Molecular Weight: 194,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08757581
CAS-Nummer: 202934-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR