Methyl 3-(1-Imidazolyl)benzoate

Lieferant: Apollo Scientific

OR470028-5G
APOSOR470028-5GEA 337 EUR
APOSOR470028-5G
Methyl 3-(1-Imidazolyl)benzoate
Methyl 3-(1-Imidazolyl)benzoate
Formel: C₁₁H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12198856
CAS-Nummer: 335255-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR