1-(4-Nitrophenyl)cyclopentanecarbonitrile
Lieferant: Apollo Scientific
OR470030-1G
APOSOR470030-1GEA
357
EUR
APOSOR470030-1G
1-(4-Nitrophenyl)cyclopentanecarbonitrile
1-(4-Nitrophenyl)cyclopentanecarbonitrile
Formel:
C₁₂H₁₂N₂O₂ Molecular Weight: 216,236 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01012887 CAS-Nummer: 91392-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...