1-Boc-4-[(6-chloro-3-pyridazinyl)amino]piperidine

Lieferant: Apollo Scientific

OR470033-5G
APOSOR470033-5GEA 499 EUR
APOSOR470033-5G
1-Boc-4-[(6-chloro-3-pyridazinyl)amino]piperidine
1-Boc-4-[(6-chloro-3-pyridazinyl)amino]piperidine
Formel: C₁₄H₂₁ClN₄O₂
Molecular Weight: 312,795 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09607520
CAS-Nummer: 939986-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR