Ethyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate

Lieferant: Apollo Scientific

OR470035-5G
APOSOR470035-5GEA 367 EUR
APOSOR470035-5G
Ethyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Ethyl 3-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Formel: C₁₃H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12198872
CAS-Nummer: 1166756-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR