Methyl 4-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate

Lieferant: Apollo Scientific

OR470036-5G
APOSOR470036-5GEA 367 EUR
APOSOR470036-5G
Methyl 4-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Methyl 4-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Formel: C₁₂H₁₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12198873
CAS-Nummer: 196301-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR