Ethyl 4-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate

Lieferant: Apollo Scientific

OR470037-5G
APOSOR470037-5GEA 367 EUR
APOSOR470037-5G
Ethyl 4-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Ethyl 4-(5-Ethyl-1,2,4-oxadiazol-3-yl)benzoate
Formel: C₁₃H₁₄N₂O₃
Molecular Weight: 246,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198874
CAS-Nummer: 1166756-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR