(3aR,6aR)-1-Methyl-5-ethoxycarbonylhexahydropyrrolo[3,4-b]pyrrole

Lieferant: Apollo Scientific

OR470040-1G
APOSOR470040-1GEA 672 EUR
APOSOR470040-1G
(3aR,6aR)-1-Methyl-5-ethoxycarbonylhexahydropyrrolo[3,4-b]pyrrole
(3aR,6aR)-1-Methyl-5-ethoxycarbonylhexahydropyrrolo[3,4-b]pyrrole
Formel: C₁₀H₁₈N₂O₂
Molecular Weight: 198,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198888
CAS-Nummer: 1417789-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR