2-Acetamido-4-methoxy-5-nitrotoluene

Lieferant: Apollo Scientific

OR470042-5G
APOSOR470042-5GEA 561 EUR
APOSOR470042-5G
2-Acetamido-4-methoxy-5-nitrotoluene
2-Acetamido-4-methoxy-5-nitrotoluene
Formel: C₁₀H₁₂N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD12198895
CAS-Nummer: 361162-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR