Ethyl 6-(1-Pyrrolidyl)pyridine-2-carboxylate

Lieferant: Apollo Scientific

OR470045-5G
APOSOR470045-5GEA 916 EUR
APOSOR470045-5G
Ethyl 6-(1-Pyrrolidyl)pyridine-2-carboxylate
Ethyl 6-(1-Pyrrolidyl)pyridine-2-carboxylate
Formel: C₁₂H₁₆N₂O₂
Molecular Weight: 220,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198900
CAS-Nummer: 1166756-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR