Ethyl 6-Morpholinopyridine-2-carboxylate

Lieferant: Apollo Scientific

OR470046-5G
APOSOR470046-5GEA 713 EUR
APOSOR470046-5G
Ethyl 6-Morpholinopyridine-2-carboxylate
Ethyl 6-Morpholinopyridine-2-carboxylate
Formel: C₁₂H₁₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12198901
CAS-Nummer: 1061750-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR