(R)-1-(1H-Benzimidazol-2-yl)-2-methylpropylamine hydrochloride

Lieferant: Apollo Scientific

OR470050-5G
APOSOR470050-5GEA 916 EUR
APOSOR470050-5G
(R)-1-(1H-Benzimidazol-2-yl)-2-methylpropylamine hydrochloride
(R)-1-(1H-Benzimidazol-2-yl)-2-methylpropylamine hydrochloride
Formel: C₁₁H₁₅N₃·ClH
Molecular Weight: 225,718 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12828691
CAS-Nummer: 1234863-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR