Ethyl 2-(Methylsulphonyl)nicotinate

Lieferant: Apollo Scientific

OR470054-5G
APOSOR470054-5GEA 804 EUR
APOSOR470054-5G
Ethyl 2-(Methylsulphonyl)nicotinate
Ethyl 2-(Methylsulphonyl)nicotinate
Formel: C₉H₁₁NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD12828713
CAS-Nummer: 1186663-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR