2-[4-(Cyclopentyloxy)phenyl]acetonitrile

Lieferant: Apollo Scientific

OR470056-5G
APOSOR470056-5GEA 601 EUR
APOSOR470056-5G
2-[4-(Cyclopentyloxy)phenyl]acetonitrile
2-[4-(Cyclopentyloxy)phenyl]acetonitrile
Formel: C₁₃H₁₅NO
Molecular Weight: 201,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08166709
CAS-Nummer: 900137-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR