3-Bromo-4-piperazinobenzoic acid
Lieferant: Apollo Scientific
OR470063-5G
APOSOR470063-5GEA
804
EUR
APOSOR470063-5G
3-Bromo-4-piperazinobenzoic acid
3-Bromo-4-piperazinobenzoic acid
Formel:
C₁₁H₁₃BrN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110935 CAS-Nummer: 1131594-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...